[(5-Methylimidazo[1,2-a]pyridin-2-yl)methyl]amine

Supplier: Matrix Scientific

039974-500MG
101848-586EA 484.62 USD
101848-586
[(5-Methylimidazo[1,2-a]pyridin-2-yl)methyl]amine
[(5-Methylimidazo[1,2-a]pyridin-2-yl)methyl]amine

Hazard: Irritant

Formula: C₉H₁₁N₃
MDL Number: MFCD08669763
CAS Number: 880361-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR