3-Chloro-6-(propylamino)pyridazine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N-propylpyridazin-3-amine

039973-500MG
101848-584EA 484.62 USD
101848-584
3-Chloro-6-(propylamino)pyridazine
3-Chloro-6-(propylamino)pyridazine

Hazard: Irritant

Formula: C₇H₁₀ClN₃
MW: 171.6274 g/mol
MDL Number: MFCD09800940
CAS Number: 951885-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR