6-Chloro-N,N-diethylpyridazin-3-amine

Supplier: Matrix Scientific

039971-500MG
101848-580EA 362.25 USD
101848-580
6-Chloro-N,N-diethylpyridazin-3-amine
6-Chloro-N,N-diethylpyridazin-3-amine

Hazard: Irritant

Formula: C₈H₁₂ClN₃
MDL Number: MFCD11205099
CAS Number: 55825-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR