Methyl-4-hydroxy-2H-thieno[2,3-e][1,2]thiazine-3-carboxylate 1,1-dioxide
Supplier: Matrix Scientific
039962-500MG
101848-552EA
193.85
USD
101848-552
Methyl-4-hydroxy-2H-thieno[2,3-e][1,2]thiazine-3-carboxylate 1,1-dioxide
Methyl-4-hydroxy-2H-thieno[2,3-e][1,2]thiazine-3-carboxylate 1,1-dioxide
Hazard: Irritant
Formula:
C₈H₇NO₅S₂ |
MDL Number:
MFCD08460399 CAS Number: 98827-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...