5-(2-Bromoethyl)pyrimidine-2,4(1H,3H)-dione

Supplier: Matrix Scientific

039960-500MG
101848-548EA 443.08 USD
101848-548
5-(2-Bromoethyl)pyrimidine-2,4(1H,3H)-dione
5-(2-Bromoethyl)pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₆H₇BrN₂O₂
MDL Number: MFCD12028113
CAS Number: 53438-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR