3-Hydroxycyclopent-2-en-1-one

Supplier: Matrix Scientific

039954-500MG
101848-536EA 110.77 USD
101848-536
3-Hydroxycyclopent-2-en-1-one
3-Hydroxycyclopent-2-en-1-one

Hazard: Irritant

Formula: C₅H₆O₂
MDL Number: MFCD12028109
CAS Number: 5870-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR