N-(2-Hydroxyethyl)-4-methylbenzamide

Supplier: Matrix Scientific

039951-500MG
101848-528EA 146.15 USD
101848-528
N-(2-Hydroxyethyl)-4-methylbenzamide
N-(2-Hydroxyethyl)-4-methylbenzamide

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00277048
CAS Number: 42469-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR