Isoxazolo[5,4-b]pyridin-3-ylamine

Supplier: Matrix Scientific

Synonyms: Isoxazolo[5,4-b]pyridin-3-amine

039947-1G
101848-518EA 507.69 USD
101848-518
Isoxazolo[5,4-b]pyridin-3-ylamine
Isoxazolo[5,4-b]pyridin-3-ylamine

Hazard: Irritant

Formula: C₆H₅N₃O
MW: 135.13 g/mol
MDL Number: MFCD04117798
CAS Number: 92914-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR