6-Chloro-N-isopropylpyridazin-3-amine

Supplier: Matrix Scientific

039938-500MG
101848-496EA 193.85 USD
101848-496
6-Chloro-N-isopropylpyridazin-3-amine
6-Chloro-N-isopropylpyridazin-3-amine

Hazard: Irritant

Formula: C₇H₁₀ClN₃
MDL Number: MFCD11125896
CAS Number: 1007-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR