3-Chloro-6-ethoxypyridazine

Supplier: Matrix Scientific

039937-500MG
101848-494EA 144.9 USD
101848-494
3-Chloro-6-ethoxypyridazine
3-Chloro-6-ethoxypyridazine

Hazard: Irritant

Formula: C₆H₇ClN₂O
MW: 158.5900 g/mol
MDL Number: MFCD00270290
CAS Number: 17321-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR