[2-(4-Pyrimidin-2-ylpiperazin-1-yl)ethyl]aminetrihydrochloride

Supplier: Matrix Scientific

039936-500MG
101848-492EA 387.69 USD
101848-492
[2-(4-Pyrimidin-2-ylpiperazin-1-yl)ethyl]aminetrihydrochloride
[2-(4-Pyrimidin-2-ylpiperazin-1-yl)ethyl]aminetrihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀Cl₃N₅
MDL Number: MFCD18071186
CAS Number: 30194-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR