1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride

Supplier: Matrix Scientific

039934-500MG
101848-488EA 150.77 USD
101848-488
1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride
1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₀Cl₂N₂O
MW: 243.18 g/mol
MDL Number: MFCD11858471
CAS Number: 790223-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR