1-(Tetrahydro-2H-pyran-4-yl)piperidine-4-carboxylic acid hydrochloride

Supplier: Matrix Scientific

039932-500MG
101848-484EA 289.8 USD
101848-484
1-(Tetrahydro-2H-pyran-4-yl)piperidine-4-carboxylic acid hydrochloride
1-(Tetrahydro-2H-pyran-4-yl)piperidine-4-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₃
MDL Number: MFCD12028102
CAS Number: 1158712-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR