Methyl (4-aminopiperidin-1-yl)acetate dihydrochloride

Supplier: Matrix Scientific

Synonyms: 4-Amino-1-piperidineacetic acid methyl ester hydrochloride

039930-500MG
101848-480EA 290.77 USD
101848-480
Methyl (4-aminopiperidin-1-yl)acetate dihydrochloride
Methyl (4-aminopiperidin-1-yl)acetate dihydrochloride

Hazard: Irritant

Formula: C₈H₁₈Cl₂N₂O₂
MW: 245.15 g/mol
MDL Number: MFCD12028101
CAS Number: 90152-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR