1-(Pyridin-3-ylmethyl)piperidin-3-aminetrihydrochloride

Supplier: Matrix Scientific

039923-500MG
101848-466EA 484.62 USD
101848-466
1-(Pyridin-3-ylmethyl)piperidin-3-aminetrihydrochloride
1-(Pyridin-3-ylmethyl)piperidin-3-aminetrihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀Cl₃N₃
MDL Number: MFCD12028099
CAS Number: 1158689-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR