N-Methyl-2-piperidin-4-ylacetamide hydrochloride

Supplier: Matrix Scientific

039922-500MG
101848-464EA 290.77 USD
101848-464
N-Methyl-2-piperidin-4-ylacetamide hydrochloride
N-Methyl-2-piperidin-4-ylacetamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O
MDL Number: MFCD12028098
CAS Number: 70724-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR