1-(2-Thienylmethyl)piperidin-4-amine

Supplier: Matrix Scientific

039921-500MG
101848-462EA 484.62 USD
101848-462
1-(2-Thienylmethyl)piperidin-4-amine
1-(2-Thienylmethyl)piperidin-4-amine

Hazard: Irritant

Formula: C₁₀H₁₆N₂S
MDL Number: MFCD10005762
CAS Number: 82378-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR