4-Isopropoxy-6-phenyl-1,3,5-triazin-2-amine

Supplier: Matrix Scientific

039920-500MG
101848-460EA 323.08 USD
101848-460
4-Isopropoxy-6-phenyl-1,3,5-triazin-2-amine
4-Isopropoxy-6-phenyl-1,3,5-triazin-2-amine

Hazard: Irritant

Formula: C₁₂H₁₄N₄O
MDL Number: MFCD00450517
CAS Number: 400839-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR