4-Methoxy-6-phenyl-1,3,5-triazin-2-amine

Supplier: Matrix Scientific

039914-500MG
101848-448EA 443.08 USD
101848-448
4-Methoxy-6-phenyl-1,3,5-triazin-2-amine
4-Methoxy-6-phenyl-1,3,5-triazin-2-amine

Hazard: Irritant

Formula: C₁₀H₁₀N₄O
MDL Number: MFCD00458219
CAS Number: 30369-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR