4-tert-Butyl-6-methoxy-1,3,5-triazin-2-amine
Supplier: Matrix Scientific
039912-500MG
101848-444EA
387.69
USD
101848-444
4-tert-Butyl-6-methoxy-1,3,5-triazin-2-amine
4-tert-Butyl-6-methoxy-1,3,5-triazin-2-amine
Hazard: Irritant
Formula:
C₈H₁₄N₄O |
MDL Number:
MFCD00052784 CAS Number: 175204-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...