5-(2-Chlorophenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

039909-500MG
101848-438EA 193.85 USD
101848-438
5-(2-Chlorophenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol
5-(2-Chlorophenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₄H₁₀ClN₃S
MDL Number: MFCD01204096
CAS Number: 23711-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR