5-tert-Butyl-4-ethyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

039906-500MG
101848-432EA 290.77 USD
101848-432
5-tert-Butyl-4-ethyl-4H-1,2,4-triazole-3-thiol
5-tert-Butyl-4-ethyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₈H₁₅N₃S
MDL Number: MFCD12028094
CAS Number: 401630-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR