5-(3-Methoxyphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

039905-500MG
101848-430EA 116.92 USD
101848-430
5-(3-Methoxyphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol
5-(3-Methoxyphenyl)-4-phenyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₅H₁₃N₃OS
MDL Number: MFCD00423358
CAS Number: 174573-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR