5-(2-Chlorophenyl)-N-ethyl-1,3,4-thiadiazol-2-amine

Supplier: Matrix Scientific

039904-500MG
101848-428EA 387.69 USD
101848-428
5-(2-Chlorophenyl)-N-ethyl-1,3,4-thiadiazol-2-amine
5-(2-Chlorophenyl)-N-ethyl-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₁₀H₁₀ClN₃S
MDL Number: MFCD01821399
CAS Number: 35313-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR