N-Allyl-5-(3-methoxyphenyl)-1,3,4-thiadiazol-2-amine
Supplier: Matrix Scientific
039900-500MG
101848-420EA
289.8
USD
101848-420
N-Allyl-5-(3-methoxyphenyl)-1,3,4-thiadiazol-2-amine
N-Allyl-5-(3-methoxyphenyl)-1,3,4-thiadiazol-2-amine
Hazard: Irritant
Formula:
C₁₂H₁₃N₃OS |
MDL Number:
MFCD00611171 CAS Number: 1134335-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...