N-Ethyl-5-phenyl-1,3,4-thiadiazol-2-amine

Supplier: Matrix Scientific

039896-500MG
101848-412EA 387.69 USD
101848-412
N-Ethyl-5-phenyl-1,3,4-thiadiazol-2-amine
N-Ethyl-5-phenyl-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₁₀H₁₁N₃S
MDL Number: MFCD01821398
CAS Number: 91265-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR