1-(4-Nitrophenyl)-1H-pyrrole-2-carbaldehyde
Supplier: Matrix Scientific
039895-500MG
101848-410EA
362.25
USD
101848-410
1-(4-Nitrophenyl)-1H-pyrrole-2-carbaldehyde
1-(4-Nitrophenyl)-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₁₁H₈N₂O₃ |
MDL Number:
MFCD00452154 CAS Number: 30186-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...