2-(5-Chloro-2-hydroxyphenyl)-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

039893-500MG
101848-406EA 387.69 USD
101848-406
2-(5-Chloro-2-hydroxyphenyl)-1H-isoindole-1,3(2H)-dione
2-(5-Chloro-2-hydroxyphenyl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₄H₈ClNO₃
MDL Number: MFCD00432185
CAS Number: 80460-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR