Ethyl-2-(4-isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)nicotinate

Supplier: Matrix Scientific

039891-500MG
101848-402EA 362.25 USD
101848-402
Ethyl-2-(4-isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)nicotinate
Ethyl-2-(4-isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)nicotinate

Hazard: Irritant

Formula: C₁₅H₁₉N₃O₃
MDL Number: MFCD00233483
CAS Number: 81333-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR