[4-(Methylthio)phenoxy]acetic acid
Supplier: Matrix Scientific
039883-500MG
101848-386EA
217.35
USD
101848-386
[4-(Methylthio)phenoxy]acetic acid
[4-(Methylthio)phenoxy]acetic acid
Hazard: Irritant
Formula:
C₉H₁₀O₃S |
MDL Number:
MFCD00233485 CAS Number: 15267-49-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...