5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-Methyl-1-phenylpyrazole-4-carboxylic acid

039881-500MG
101848-382EA 289.8 USD
101848-382
5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid
5-Methyl-1-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD00067831
CAS Number: 91138-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR