3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid

Supplier: Matrix Scientific

039878-500MG
101848-376EA 289.8 USD
101848-376
3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid
3,5-Dimethyl-1-phenyl-1H-pyrazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD02656670
CAS Number: 61226-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR