(1,4,5,6,7,8-Hexahydrocyclohepta[c]pyrazol-3-ylmethyl)amine hydrochloride

Supplier: Matrix Scientific

039877-500MG
101848-374EA 217.35 USD
101848-374
(1,4,5,6,7,8-Hexahydrocyclohepta[c]pyrazol-3-ylmethyl)amine hydrochloride
(1,4,5,6,7,8-Hexahydrocyclohepta[c]pyrazol-3-ylmethyl)amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClN₃
MDL Number: MFCD12028091
CAS Number: 1185302-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR