Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1,4,5,6-Tetrahydrocyclopenta[c]pyrazol-3-ylmethyl)amine hydrochloride

Supplier: Matrix Scientific


039876-500MG
101848-372EA 411.92 USD InStock
101848-372
(1,4,5,6-Tetrahydrocyclopenta[c]pyrazol-3-ylmethyl)amine hydrochloride
(1,4,5,6-Tetrahydrocyclopenta[c]pyrazol-3-ylmethyl)amine hydrochloride
CAS: 1185295-94-6
MDL Number: MFCD12028090
Molecular Formula: C7H12ClN3
Molecular Weight: 173,65
Hazard: Irritant


Formula: C₇H₁₂ClN₃
MW: 173,65 g/mol
MDL Number: MFCD12028090
CAS Number: 1185295-94-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty