Ethyl-2-(1H-benzimidazol-2-ylthio)propanoate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-((1H-benzo[d]imidazol-2-yl)thio)propanoate

039875-500MG
101848-370EA 217.35 USD
101848-370
Ethyl-2-(1H-benzimidazol-2-ylthio)propanoate
Ethyl-2-(1H-benzimidazol-2-ylthio)propanoate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂S
MW: 250.32 g/mol
MDL Number: MFCD00521808
CAS Number: 160004-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR