2-Chloro-3-(5-isopropyl-1,2,4-oxadiazol-3-yl)-6-methylquinoline
Supplier: Matrix Scientific
039871-500MG
101848-362EA
289.8
USD
101848-362
2-Chloro-3-(5-isopropyl-1,2,4-oxadiazol-3-yl)-6-methylquinoline
2-Chloro-3-(5-isopropyl-1,2,4-oxadiazol-3-yl)-6-methylquinoline
Hazard: Irritant
Formula:
C₁₅H₁₄ClN₃O |
MDL Number:
MFCD12028087 CAS Number: 1142201-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...