2-Chloro-7-methyl-3-(5-methyl-1,2,4-oxadiazol-3-yl)quinoline

Supplier: Matrix Scientific

039870-500MG
101848-360EA 387.69 USD
101848-360
2-Chloro-7-methyl-3-(5-methyl-1,2,4-oxadiazol-3-yl)quinoline
2-Chloro-7-methyl-3-(5-methyl-1,2,4-oxadiazol-3-yl)quinoline

Hazard: Irritant

Formula: C₁₃H₁₀ClN₃O
MDL Number: MFCD12028086
CAS Number: 1142201-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR