2-Chloro-6-methyl-3-(5-propyl-1,2,4-oxadiazol-3-yl)quinoline

Supplier: Matrix Scientific

039868-500MG
101848-356EA 387.69 USD
101848-356
2-Chloro-6-methyl-3-(5-propyl-1,2,4-oxadiazol-3-yl)quinoline
2-Chloro-6-methyl-3-(5-propyl-1,2,4-oxadiazol-3-yl)quinoline

Hazard: Irritant

Formula: C₁₅H₁₄ClN₃O
MDL Number: MFCD12028084
CAS Number: 1142201-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR