2-Chloro-8-methyl-3-(5-phenyl-1,2,4-oxadiazol-3-yl)quinoline

Supplier: Matrix Scientific

039866-500MG
101848-352EA 290.77 USD
101848-352
2-Chloro-8-methyl-3-(5-phenyl-1,2,4-oxadiazol-3-yl)quinoline
2-Chloro-8-methyl-3-(5-phenyl-1,2,4-oxadiazol-3-yl)quinoline

Hazard: Irritant

Formula: C₁₈H₁₂ClN₃O
MDL Number: MFCD12028082
CAS Number: 1142201-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR