2-Chloro-7-methyl-3-(5-propyl-1,2,4-oxadiazol-3-yl)quinoline
Supplier: Matrix Scientific
039865-500MG
101848-350EA
290.77
USD
101848-350
2-Chloro-7-methyl-3-(5-propyl-1,2,4-oxadiazol-3-yl)quinoline
2-Chloro-7-methyl-3-(5-propyl-1,2,4-oxadiazol-3-yl)quinoline
Hazard: Irritant
Formula:
C₁₅H₁₄ClN₃O |
MDL Number:
MFCD12028081 CAS Number: 1142201-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...