3-Bromo-4-methylquinolin-2(1H)-one

Supplier: Matrix Scientific

039864-500MG
101848-348EA 443.08 USD
101848-348
3-Bromo-4-methylquinolin-2(1H)-one
3-Bromo-4-methylquinolin-2(1H)-one

Hazard: Irritant

Formula: C₁₀H₈BrNO
MDL Number: MFCD01740154
CAS Number: 37778-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR