2-Methylbutane-1,4-diamine dihydrochloride

Supplier: Matrix Scientific

039861-500MG
101848-342EA 362.25 USD
101848-342
2-Methylbutane-1,4-diamine dihydrochloride
2-Methylbutane-1,4-diamine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₆Cl₂N₂
MDL Number: MFCD12028080
CAS Number: 111469-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR