2-(Trifluoromethyl)-4H-3,1-benzoxazin-4-one

Supplier: Matrix Scientific

039860-500MG
101848-340EA 290.77 USD
101848-340
2-(Trifluoromethyl)-4H-3,1-benzoxazin-4-one
2-(Trifluoromethyl)-4H-3,1-benzoxazin-4-one

Hazard: Irritant

Formula: C₉H₄F₃NO₂
MDL Number: MFCD00175074
CAS Number: 16062-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR