2,6-Diethyl-3,5-dimethyl-4H-pyran-4-one

Supplier: Matrix Scientific

039858-500MG
101848-334EA 384.62 USD
101848-334
2,6-Diethyl-3,5-dimethyl-4H-pyran-4-one
2,6-Diethyl-3,5-dimethyl-4H-pyran-4-one

Hazard: Irritant

Formula: C₁₁H₁₆O₂
MDL Number: MFCD01366758
CAS Number: 1202-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR