6-Iodo-2H-3,1-benzoxazine-2,4(1H)-dione

Supplier: Matrix Scientific

Synonyms: 6-Iodo-1H-benzo[d][1,3]oxazine-2,4-dione

039855-500MG
101848-328EA 289.8 USD
101848-328
6-Iodo-2H-3,1-benzoxazine-2,4(1H)-dione
6-Iodo-2H-3,1-benzoxazine-2,4(1H)-dione

Hazard: Irritant

Formula: C₈H₄INO₃
MW: 289.03 g/mol
MDL Number: MFCD06661932
CAS Number: 116027-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR