1-(4-Nitrophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039852-500MG
101848-320EA 290.77 USD
101848-320
1-(4-Nitrophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-(4-Nitrophenyl)-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₆ClN₃O₂
MW: 329.79 g/mol
MDL Number: MFCD07626284
CAS Number: 3380-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR