1-(4-Chlorophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039850-500MG
101848-316EA 290.77 USD
101848-316
1-(4-Chlorophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-(4-Chlorophenyl)-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₆Cl₂N₂
MW: 319.24 g/mol
MDL Number: MFCD01690472
CAS Number: 3380-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR