1-(4-Chloro-3-nitrophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039849-500MG
101848-314EA 290.77 USD
101848-314
1-(4-Chloro-3-nitrophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-(4-Chloro-3-nitrophenyl)-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₅Cl₂N₃O₂
MW: 364.23 g/mol
MDL Number: MFCD01690470
CAS Number: 3380-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR