1-(3-Nitrophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride

Supplier: Matrix Scientific

039848-500MG
101848-312EA 290.77 USD
101848-312
1-(3-Nitrophenyl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-(3-Nitrophenyl)-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₆ClN₃O₂
MW: 329.79 g/mol
MDL Number: MFCD01749633
CAS Number: 102207-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR