1-(1,3-Benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
Supplier: Matrix Scientific
039847-500MG
101848-310EA
387.69
USD
101848-310
1-(1,3-Benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-β-carboline hydrochloride
1-(1,3-Benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-beta-carboline hydrochloride
Hazard: Irritant
Formula:
C₁₈H₁₇ClN₂O₂ MW: 328.8 g/mol |
MDL Number:
MFCD00817133 CAS Number: 3380-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...